Learn More
Stoelting™ T Maze
SDP
Catalog No. 10000307 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Rat
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal
10-000-307 Mouse
10-000-330 Rat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-307 Supplier Stoelting™ Supplier No. 60181
Only null left
Add to Cart
Add to Cart
Stoelting™ T-maze is used largely in preference and spatial learning tasks.
Animals learn to alternate between arms based on their memory of the previously visited arms or choose an arm based on the reward presented. The simple capital T shape design incorporates a single choice point with only two alternatives.
Specifications
For Use With (Application) Neuroscience
Animal Mouse