Learn More
MSC T-NUT MOQ 15 EACH
Catalog No. NC2945711
Encompass
This bolt assembly is designed for fastening applications within structural systems. It includes a flanged button head socket cap screw and a slide-in economy T-nut with anP offset thread.
  • Flanged button head socket cap screw
  • Slide-in economy T-nut with offset thread
  • Thread size: 5/16-18
  • Black zinc finish
Catalog No. NC2945711 Supplier MSC Supplier No. 07162944
This bolt assembly is designed for fastening applications within structural systems. It includes a flanged button head socket cap screw and a slide-in economy T-nut with anP offset thread.
  • Flanged button head socket cap screw
  • Slide-in economy T-nut with offset thread
  • Thread size: 5/16-18
  • Black zinc finish