Learn More
Grainger T-Nut with Ball Spring 80/20 15 Series
Catalog No. 502767452
Encompass_Preferred
The 80/20 T-Nut 15 Series is a T-slot nut designed for use with 80/20 15 series aluminum extrusion profiles. It is made from zinc-plated steel for corrosion resistance and has dimensions of 0.625" width x 0.375" height x 0.375" depth. The T-nut features a center rib that slides into the T-slot on 15 series extrusions and allows the nut to be securely fastened anywhere along the length of the profile using an M5 screw. This versatile T-nut provides a simple way to attach components to 80/20 15 series aluminum framing.
Catalog No. 50-276-7452 Supplier Grainger Supplier No. 16U347
The 80/20 T-Nut 15 Series is a T-slot nut designed for use with 80/20 15 series aluminum extrusion profiles. It is made from zinc-plated steel for corrosion resistance and has dimensions of 0.625" width x 0.375" height x 0.375" depth. The T-nut features a center rib that slides into the T-slot on 15 series extrusions and allows the nut to be securely fastened anywhere along the length of the profile using an M5 screw. This versatile T-nut provides a simple way to attach components to 80/20 15 series aluminum framing.