Learn More
Medchemexpress LLC T-PA Human HEK293 100ug
SDP
Catalog No. 5000196869
Encompass_Preferred
Recombinant human tissue plasminogen activator (t-PA) expressed in HEK293 cells with a C-terminal His tag. Supplied as a lyophilized powder with low endotoxin and high specific activity, it is suitable for enzymatic assays, fibrinolysis research, and biochemical characterization when stored and handled according to manufacturer recommendations.
  • Human-sequence protein expressed in HEK293 cells.
  • His tag facilitates purification and detection.
  • Lyophilized formulation for stable storage and transport.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • High enzymatic activity (>500.47 pmol/min/μg) for functional studies.
  • Recommended storage at -20°C; aliquot and freeze at -20°C or -80°C for long-term use.
Catalog No. 50-001-96869 Supplier Medchemexpress LLC Supplier No. HYP71051100UG
Recombinant human tissue plasminogen activator (t-PA) expressed in HEK293 cells with a C-terminal His tag. Supplied as a lyophilized powder with low endotoxin and high specific activity, it is suitable for enzymatic assays, fibrinolysis research, and biochemical characterization when stored and handled according to manufacturer recommendations.
  • Human-sequence protein expressed in HEK293 cells.
  • His tag facilitates purification and detection.
  • Lyophilized formulation for stable storage and transport.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • High enzymatic activity (>500.47 pmol/min/μg) for functional studies.
  • Recommended storage at -20°C; aliquot and freeze at -20°C or -80°C for long-term use.