Learn More
Medchemexpress LLC T-PA Human HEK293 10ug
SDP
Catalog No. 5000180505
Encompass_Preferred
The T-PA protein encoded by this gene is a secreted serine protease that converts plasminogen into plasmin a key enzyme involved in tissue remodeling and cell migration The proteolytic process produces heavy and light chains forming heterodimeric enzymes T-PA Protein Human (HEK293) is the recombinant human-derived T-PA protein expressed by HEK293 with tag free
Catalog No. 50-001-80505 Supplier Medchemexpress LLC Supplier No. HYP7345110UG
The T-PA protein encoded by this gene is a secreted serine protease that converts plasminogen into plasmin a key enzyme involved in tissue remodeling and cell migration The proteolytic process produces heavy and light chains forming heterodimeric enzymes T-PA Protein Human (HEK293) is the recombinant human-derived T-PA protein expressed by HEK293 with tag free