Learn More
Medchemexpress LLC T-PA Human HEK293 5ug
SDP
Catalog No. 5000197110
Encompass_Preferred
Recombinant human tissue plasminogen activator (tPA) produced in HEK293 cells with a C-terminal His tag. Supplied at high purity and suitable for biochemical and fibrinolysis assays, the glycosylated protein has an apparent molecular weight of approximately 62-72 kDa and is provided in a 5 μg quantity for research applications.
  • Expressed in HEK293 cells.
  • Contains a C-terminal His tag for affinity purification.
  • High purity suitable for biochemical assays (~98%).
  • Glycosylated protein with apparent molecular weight ~62-72 kDa.
  • Supplied as a 5 μg quantity for small-scale studies.
  • For research use only.
Catalog No. 50-001-97110 Supplier Medchemexpress LLC Supplier No. HYP710515UG
Recombinant human tissue plasminogen activator (tPA) produced in HEK293 cells with a C-terminal His tag. Supplied at high purity and suitable for biochemical and fibrinolysis assays, the glycosylated protein has an apparent molecular weight of approximately 62-72 kDa and is provided in a 5 μg quantity for research applications.
  • Expressed in HEK293 cells.
  • Contains a C-terminal His tag for affinity purification.
  • High purity suitable for biochemical assays (~98%).
  • Glycosylated protein with apparent molecular weight ~62-72 kDa.
  • Supplied as a 5 μg quantity for small-scale studies.
  • For research use only.