Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tissue-type plasminogen activator (mouse) | >95.0% | 50UG
SDP

Catalog No. 5000174947
Encompass_Preferred

Recombinant tissue-type plasminogen activator (mouse) produced in HEK293 cells with a C-terminal His tag, supplied as a lyophilized powder for research use in plasminogen activation and fibrinolysis assays. The material is quality-tested for purity and endotoxin and shows multiple glycosylated bands on SDS-PAGE.

  • Recombinant mouse tissue-type plasminogen activator expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Purity greater than 95.0% by Tris-Bis PAGE.
  • Apparent molecular weight ~13-15 kDa, ~28-34 kDa, and ~35-40 kDa due to glycosylation.
  • Endotoxin level <1 EU/μg as determined by LAL.
  • Supplied as lyophilized powder; reconstitute at ≥100 μg/mL in ddH2O.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or -20°C longer with carrier protein; aliquot for long-term storage.

Catalog No. 50-001-74947 Supplier Medchemexpress LLC Supplier No. HYP70131250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant tissue-type plasminogen activator (mouse) produced in HEK293 cells with a C-terminal His tag, supplied as a lyophilized powder for research use in plasminogen activation and fibrinolysis assays. The material is quality-tested for purity and endotoxin and shows multiple glycosylated bands on SDS-PAGE.

  • Recombinant mouse tissue-type plasminogen activator expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Purity greater than 95.0% by Tris-Bis PAGE.
  • Apparent molecular weight ~13-15 kDa, ~28-34 kDa, and ~35-40 kDa due to glycosylation.
  • Endotoxin level <1 EU/μg as determined by LAL.
  • Supplied as lyophilized powder; reconstitute at ≥100 μg/mL in ddH2O.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or -20°C longer with carrier protein; aliquot for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.