Learn More
Medchemexpress LLC T-PA Mouse HEK293 5ug
SDP
Catalog No. 5000180574
Encompass_Preferred
The T-PA protein converts the inactive plasminogen to active plasmin by hydrolyzing a single Arg-Val bond This conversion regulates plasmin-mediated proteolysis and is involved in tissue remodeling degradation and cell migration T-PA also contributes to the prevention of polyspermy during oocyte activation by participating in the cortical granule reaction T-PA Protein Mouse (HEK293 Fc) is the recombinant mouse-derived T-PA protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-80574 Supplier Medchemexpress LLC Supplier No. HYP735835UG
The T-PA protein converts the inactive plasminogen to active plasmin by hydrolyzing a single Arg-Val bond This conversion regulates plasmin-mediated proteolysis and is involved in tissue remodeling degradation and cell migration T-PA also contributes to the prevention of polyspermy during oocyte activation by participating in the cortical granule reaction T-PA Protein Mouse (HEK293 Fc) is the recombinant mouse-derived T-PA protein expressed by HEK293 with N-hFc labeled tag