Learn More
Medchemexpress LLC T-PA protein, mouse (HEK293, His) | >95.0% | 5UG
SDP
Supplier:  Medchemexpress LLC HYP7013125UG
Encompass_Preferred
Recombinant mouse tissue-type plasminogen activator (tPA) expressed in HEK293 cells with a C-terminal His tag. Supplied as a purified protein for research use in biochemical and cell-based assays involving plasminogen activation and fibrinolysis. Multiple bands on SDS-PAGE reflect glycosylation.
Catalog No. 50-001-76718