Learn More
Creative Biomart Recombinant T.pallidum p15 Antigen, b-galactosidase-tagged
Catalog No. 502084804
Encompass_Preferred
The E. coli derived recombinant protein contains the Trp. Pallidum p15 immunodominant regions. The protein contains beta-galactosidase (114 kDa) fused at the N-terminus.
Catalog No. 50-208-4804 Supplier Creative Biomart Supplier No. TPP426
The E. coli derived recombinant protein contains the Trp. Pallidum p15 immunodominant regions. The protein contains beta-galactosidase (114 kDa) fused at the N-terminus.