Learn More
Creative Biomart Recombinant T.pallidum Partial p15 Antigen, His-tagged
Catalog No. 502084806
Encompass_Preferred
The E. coli derived recombinant 6 His tag fusion protein is a multimer having a molecular mass of 15 kDa and contains the Trp. Pallidum p15 immunodominant regions and six histidines fused at the C-
Catalog No. 50-208-4806 Supplier Creative Biomart Supplier No. TPP428
The E. coli derived recombinant 6 His tag fusion protein is a multimer having a molecular mass of 15 kDa and contains the Trp. Pallidum p15 immunodominant regions and six histidines fused at the C-