Learn More
Creative Biomart Recombinant T.pallidum p41 Mosaic Antigen, GST-tagged
Catalog No. 502084810
Encompass_Preferred
The E. coli derived recombinant protein contains the outer membrane T.Pallidum p41 immunodominant regions. The protein is fused with a GST tag.
Catalog No. 50-208-4810 Supplier Creative Biomart Supplier No. TPP432
The E. coli derived recombinant protein contains the outer membrane T.Pallidum p41 immunodominant regions. The protein is fused with a GST tag.