Learn More
Creative Biomart Recombinant T.pallidum p47 Antigen, b-galactosidase-tagged
Catalog No. 502084811
Encompass_Preferred
The E. coli derived recombinant protein contains the T.Pallidum p47 immunodominant regions. The protein contains beta-galactosidase (114 kDa) fused at the N-terminus.
Catalog No. 50-208-4811 Supplier Creative Biomart Supplier No. TPP433
The E. coli derived recombinant protein contains the T.Pallidum p47 immunodominant regions. The protein contains beta-galactosidase (114 kDa) fused at the N-terminus.