Learn More
Biocare Medical, LLC Treponema Pallidum Antibody - Concentrate, 0.5 ml
SDP
Supplier:  Biocare Medical, LLC ACA135B
Encompass_Preferred
Spirochete (Treponema pallidum) is the causative agent of syphilis. Spirochete can now be successfully localized with a treponema pallidum antibody using immunohistochemical techniques in formalin-fixed, paraffin-embedded (FFPE) tissue. In the past, localization of the spirochete agent was achieved with silver stains such as Steiner and/or Warthin-Starry. The spirochete antibody consists of a rabbit purified IgG fraction that is highly specific. Treponema pallidum antibody also cross-reacts with burgdorferi (Lyme disease).
Catalog No. 50-827-66