Learn More
Creative Biomart Recombinant T.pallidum Partial TmpA Antigen
Catalog No. 502084814
Encompass_Preferred
The E. coli derived 68 kDa recombinant protein contains the Trp. Pallidum TmpA immunodominant regions, 23-41 amino acids and 288-325 amino acids.
Catalog No. 50-208-4814 Supplier Creative Biomart Supplier No. TMP436
The E. coli derived 68 kDa recombinant protein contains the Trp. Pallidum TmpA immunodominant regions, 23-41 amino acids and 288-325 amino acids.