Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ T Phosphoramidite, standard grade, serum vial bottle

Catalog No. p-7182889
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC9052690 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC9052690 Supplier Thermo Scientific™ Supplier No. 27173605
Only null left
Add to Cart
Add to Cart

T CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxythymidine (dT) for use in oligonucleotide synthesis.

T CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxythymidine (dT) for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups (except T and U) designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 20 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-deoxythymidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C40H49N4O8P
Base: T
Formula weight: 744.82 g/mol
CAS number: 98796-51-1
HPLC Purity: ≥98.0%
31P NMR Purity: ≥98.0%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type T CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Deoxythymidine (dT)
Shipping Condition Room Temperature
Purity 98%
Quantity 5 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type DNA
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.