Learn More
Med Vet International "T"-Plate, 2.7mm, 5 hole, 32mm L, J0428D
Catalog No. 5000582973
Encompass_Preferred
Available for 3.5, 2.7, and 2.0mm screws, either cortical or self-tapping. "T" plates are used almost exclusively for the repair of fractures of the distal radius. In addition, they also may be used in a partial carpal arthrodesis.
Catalog No. 50-005-82973 Supplier Med Vet International Supplier No. JOR428D
Available for 3.5, 2.7, and 2.0mm screws, either cortical or self-tapping. "T" plates are used almost exclusively for the repair of fractures of the distal radius. In addition, they also may be used in a partial carpal arthrodesis.