Learn More
Med Vet International "T"-Plate, 2mm, 4 hole Right Angled, 18mm L, J0428C
Catalog No. 5000582678
Encompass_Preferred
Available for 3.5, 2.7, and 2.0mm screws, either cortical or self-tapping. "T" plates are used almost exclusively for the repair of fractures of the distal radius. In addition, they also may be used in a partial carpal arthrodesis.
Catalog No. 50-005-82678 Supplier Med Vet International Supplier No. JOR428C
Available for 3.5, 2.7, and 2.0mm screws, either cortical or self-tapping. "T" plates are used almost exclusively for the repair of fractures of the distal radius. In addition, they also may be used in a partial carpal arthrodesis.