Learn More
CPC Scientific H-Thr-Arg-Asn-Tyr-Tyr-Val-Arg-Ala-Val-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific AOXP003A
Encompass
ONE-LETTER SEQUENCE: TRNYYVRAVLMOLECULAR FORMULA: C57H91N17O15MOLECULAR WEIGHT:1254.46STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: M.-Y.Park et al., Biochem. Biophys. Res. Commun., 204, 924 (1994)
Catalog No. 50-210-8774