Learn More
Invitrogen™ T-REx™-HeLa Cell Line
Catalog No. r71407
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 107 cells
Catalog No. Quantity
R71407 1 x 107 cells
1 options
Catalog No. R71407 Supplier Invitrogen™ Supplier No. R71407
Only null left
Add to Cart
Add to Cart
Stably express the tetracycline repressor protein
T-REx™ Cell Lines stably express the tetracycline repressor protein (Table 1). They save significant time and effort when using the T-REx™ System. The T-REx™ Cell Lines are functionally tested by transient transfection with the positive control vector pcDNA™4⁄TO⁄lacZ. T-REx™ Cell Lines exhibit extremely low basal expression levels in the repressed state and high expression upon induction with tetracycline (Figure 1).
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 1 x 107 cells are supplied frozen in 1 ml 90% complete medium and 10% DMSO. Store in liquid nitrogen. Cells are guaranteed stable for 6 months when properly stored.
Cell Line T-REx-HeLa Cell Line
Species Human
No. of Cells 1 x 107
Product Line T-REx
Quantity 1 x 107 cells
WARNING: Cancer - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.