Learn More
Cosmo Bio Usa Inc T-Select I-Ab MOG 35-55 Tetramer-APC, 20 tests, Peptide Sequence MEVGWYRSPFSRVVHLYRNGK,
SDP
Catalog No. NC3433102
Encompass
MHC Tetramers are used for the detection of antigen-specific T-cell populations. MHC-peptide tetramer complexes recognize antigen specific T-cells at the single cell level, allowing for accurate measurements of the functional immune response in cancer, infectious diseases, and autoimmune diseases. Due to their increased avidity, MHC tetramers stably bind to cognate TcRs and are evaluated using flow cytometry.
Catalog No. NC3433102 Supplier Cosmo Bio Usa Inc Supplier No. MBLTSM7042
MHC Tetramers are used for the detection of antigen-specific T-cell populations. MHC-peptide tetramer complexes recognize antigen specific T-cells at the single cell level, allowing for accurate measurements of the functional immune response in cancer, infectious diseases, and autoimmune diseases. Due to their increased avidity, MHC tetramers stably bind to cognate TcRs and are evaluated using flow cytometry.