Learn More
Hampshire Controls T SENTRY MODEL 140-100H
Catalog No. NC0630710
Encompass_Preferred
User-programmable, local temperature alarm operates from-100 to +100 deg.C. Includes high & low temperature alarms, audible and visual alarm alarms, mute & delay periods. Used for monitoring freezers, refrigerators, walk-in coolers and server rooms.
Catalog No. NC0630710 Supplier Hampshire Controls Supplier No. 140100H
User-programmable, local temperature alarm operates from-100 to +100 deg.C. Includes high & low temperature alarms, audible and visual alarm alarms, mute & delay periods. Used for monitoring freezers, refrigerators, walk-in coolers and server rooms.