Learn More
Cytiva T-series Centramate™ Cassette, Omega PES membrane, 1 kDa molecular weight cut-off (MWCO), 0.02 m² effective filtration area (EFA)
Supplier:  Cytiva OS001T02
Encompass
T-series Centramate¿ Cassette, Omega PES membrane, 1 kDa molecular weight cut-off (MWCO), 0.02 m¿ effective filtration area (EFA)
Catalog No. NC2040716