Learn More
Fisherbrand™ T-Shape Disposable Ear Plugs
Catalog No. 12920003 Shop All Fisherbrand Products
Change view
Click to view available options
Corded:
Corded
Uncorded
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Corded
12-920-003 Uncorded
12-920-004 Corded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-920-003
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
T-Shape disposable ear plugs provide protection from loud noises.
  • Foam Earplugs with noise reduction
  • Non-allergenic foam product
  • Non-toxic
  • Non-irritating, with the smooth outer comfort surface
  • Protect the user's ears from loud noises
  • Individually packaged pairs, 100 pairs per box, 1000 pairs per case
Specifications
ANSI S3.1974 Yes
Size Universal
Quantity 100 Pairs/Box
Noise Reduction Rating (NRR) 31 dB
Corded Uncorded
Plug Shape T-Shape
Disposable Single-use
Metal Detectable No
Dielectric No
Mounting None
EN 352.2002 EN 352-2
EN 352 Additional Requirements EN 352-2:2020