Learn More
Alkali Scientific T-shape Spreader, 34 x 140mm, Individually Wrapped, 500/cs
SDP
Catalog No. 502389004
Encompass_Preferred
These T-shape spreaders ensures that consistently even pressure can be applied across the entire span of the 34mm spreading surface. Designed for spreading and dispersing liquids across the surface of agar culture plates, they are made of ABS and commonly used for CFU plate counts on water, milk, urine, etc.
Catalog No. 50-238-9004 Supplier Alkali Scientific Supplier No. TS5430
These T-shape spreaders ensures that consistently even pressure can be applied across the entire span of the 34mm spreading surface. Designed for spreading and dispersing liquids across the surface of agar culture plates, they are made of ABS and commonly used for CFU plate counts on water, milk, urine, etc.