Learn More
Grainger T-Shirt Black Sided Class 2 Orange 5XL
Catalog No. 502752962
Encompass_Preferred
The Grainger 14F537 is a Westward 3/8-inch drive deep impact socket set. This 15-piece set includes sizes ranging from 4mm to 14mm as well as a 3-inch extension. The sockets are made from chrome vanadium steel for durability and have a black phosphate finish to resist corrosion. The sockets feature a 6-point design that grips fasteners better than 12-point sockets. This set comes packed in a molded case for convenient storage and organization. The Westward sockets meet or exceed ANSI standards.
Catalog No. 50-275-2962 Supplier Grainger Supplier No. 14F537
The Grainger 14F537 is a Westward 3/8-inch drive deep impact socket set. This 15-piece set includes sizes ranging from 4mm to 14mm as well as a 3-inch extension. The sockets are made from chrome vanadium steel for durability and have a black phosphate finish to resist corrosion. The sockets feature a 6-point design that grips fasteners better than 12-point sockets. This set comes packed in a molded case for convenient storage and organization. The Westward sockets meet or exceed ANSI standards.