Learn More
Grainger T-Shirt Black Sided Class 3 Lime 2XL
Catalog No. 502752101
Encompass_Preferred
The Grainger Approved T-Shirt is a black, short sleeve t-shirt made of 100% cotton with a crew neckline. It has a class 3 ANSI 107 high visibility rating with 2-inch silver reflective tape around the sleeves and torso. The shirt is tag-free with a taped neck and shoulder seams. It comes in sizes small through 4XL. This t-shirt meets OSHA 1910.135 standards for high visibility safety apparel.
Catalog No. 50-275-2101 Supplier Grainger Supplier No. 14F541
The Grainger Approved T-Shirt is a black, short sleeve t-shirt made of 100% cotton with a crew neckline. It has a class 3 ANSI 107 high visibility rating with 2-inch silver reflective tape around the sleeves and torso. The shirt is tag-free with a taped neck and shoulder seams. It comes in sizes small through 4XL. This t-shirt meets OSHA 1910.135 standards for high visibility safety apparel.