Learn More
Grainger T-Shirt Black Sided Class 3 Orange 4XL
Catalog No. 502750081
Encompass_Preferred
The Grainger 14F550 is a 14-inch diameter, 550 CFM, direct drive upblast roof exhaust fan. It features a 1/6 HP, 115V, 1-phase, 60Hz motor with permanently lubricated ball bearings and automatic thermal overload protection. The housing is constructed of heavy gauge aluminum with a spun aluminum venturi for maximum airflow. It has a built-in backdraft damper and is ETL listed. This fan is ideal for exhausting heat, fumes, and vapors in commercial and industrial settings. With a max temperature rating of 104°F, it is suitable for roof or sidewall mounting.
Catalog No. 50-275-0081 Supplier Grainger Supplier No. 14F550
The Grainger 14F550 is a 14-inch diameter, 550 CFM, direct drive upblast roof exhaust fan. It features a 1/6 HP, 115V, 1-phase, 60Hz motor with permanently lubricated ball bearings and automatic thermal overload protection. The housing is constructed of heavy gauge aluminum with a spun aluminum venturi for maximum airflow. It has a built-in backdraft damper and is ETL listed. This fan is ideal for exhausting heat, fumes, and vapors in commercial and industrial settings. With a max temperature rating of 104°F, it is suitable for roof or sidewall mounting.