Learn More
MSC 80/20 Inc. T-Slotted Aluminum Extrusion, 15 Series, 1.5 in. x 1.5 in., 72 in., 1 ea.
Catalog No. 5000514601
Encompass
This 15 series lite fractional square T-slot profile is designed for structural versatility and durability. Measuring 1.5 in. x 1.5 in. with a length of 72 in., this aluminum extrusion features four open T-slots, one on each face, providing multidirectional access for mounting accessories in various frame configurations.
  • Features a smooth profile that resists dirt and debris accumulation for easy maintenance
  • Compatible with all 15 series fasteners to streamline assembly
  • Constructed from aluminum with a clear anodized finish for enhanced resistance
  • Suitable for clean room applications, guards, enclosures, and workstation frames
  • Provides efficient access from any direction for simplified accessory integration
Catalog No. 50-005-14601 Supplier MSC Supplier No. 61984886
This 15 series lite fractional square T-slot profile is designed for structural versatility and durability. Measuring 1.5 in. x 1.5 in. with a length of 72 in., this aluminum extrusion features four open T-slots, one on each face, providing multidirectional access for mounting accessories in various frame configurations.
  • Features a smooth profile that resists dirt and debris accumulation for easy maintenance
  • Compatible with all 15 series fasteners to streamline assembly
  • Constructed from aluminum with a clear anodized finish for enhanced resistance
  • Suitable for clean room applications, guards, enclosures, and workstation frames
  • Provides efficient access from any direction for simplified accessory integration