Learn More
Andwin Scientific Tough-Spots, 3/8'', Pink, 1000/roll
SDP
Catalog No. NC1663646
Encompass_Preferred
Chemically inert, polyolefin labels resist most organic solvents and caustic agents. They adhere to most plastics, glass, and metals without cracking, peeling, or degrading.
Catalog No. NC1663646 Supplier Andwin Scientific Supplier No. TSPOTSP
Chemically inert, polyolefin labels resist most organic solvents and caustic agents. They adhere to most plastics, glass, and metals without cracking, peeling, or degrading.