Learn More
Andwin Scientific T-SPOTS RED 1000/ROLL PKG
SDP
Catalog No. NC9671235
Encompass_Preferred
The red Tough-Spots labels on a sheet are specially designed to be used for 0.5 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 3/8".
Catalog No. NC9671235 Supplier Andwin Scientific Supplier No. TSPOTSR
The red Tough-Spots labels on a sheet are specially designed to be used for 0.5 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 3/8".