Learn More
Boca Scientific Inc SK-2S DNA/RNA Buccal Swab with 2ml collection tube and cap, pack of 100
Supplier:  Boca Scientific Inc SK2S
Encompass
These buccal swabs have been specifically designed to offer increased yields of high quality buccal cell and genomic DNA and all species of RNA. Each SK-2S buccal swab is packaged with a 2ml microtube and special cap. After the buccal sample is obtained, the swab is inserted in the microtube and the special cap is slid down the shaft and applied to the tube. Buccal cell sampling offers a viable alternative to blood collection and enables high yields of pure, intact DNA/RNA to be collected easily and quickly. These swabs are suitable for both human and veterinary use and offer significant advantages compared to other swab designs in terms of efficiency of cell collection by the unique swab matrix, which in conjunction with a quick release surface maximizes DNA/RNA yields. Swab batches are ethylene oxide treated and routinely tested for human DNA contamination.
Catalog No. NC9679349