Learn More
Boca Scientific Inc SK-1S DNA/RNA Buccal Swab with 5ml collection tube and cap, pack of 100
Catalog No. NC0218174
Encompass
These buccal swabs have been specifically designed to offer increased yields of high quality buccal cell and genomic DNA and all species of RNA. Each SK-1S buccal swab is packaged with a 5ml plastic collection tube and cap. After the buccal sample is obtained, the swab is inserted in the tube and the shaft is broken off just above the swab head by gentle pressure at the score. The tube is then capped and ready to be transported to the lab for processing. Buccal cell sampling offers a viable alternative to blood collection and enables high yields of pure, intact DNA/RNA to be collected easily and quickly. These swabs are suitable for both human and veterinary use and offer significant advantages compared to other swab designs in terms of efficiency of cell collection by the unique swab matrix, which in conjunction with a quick release surface maximizes DNA/RNA yields. Swab batches are ethylene oxide treated and routinely tested for human DNA contamination.
Catalog No. NC0218174 Supplier Boca Scientific Inc Supplier No. SK1S
These buccal swabs have been specifically designed to offer increased yields of high quality buccal cell and genomic DNA and all species of RNA. Each SK-1S buccal swab is packaged with a 5ml plastic collection tube and cap. After the buccal sample is obtained, the swab is inserted in the tube and the shaft is broken off just above the swab head by gentle pressure at the score. The tube is then capped and ready to be transported to the lab for processing. Buccal cell sampling offers a viable alternative to blood collection and enables high yields of pure, intact DNA/RNA to be collected easily and quickly. These swabs are suitable for both human and veterinary use and offer significant advantages compared to other swab designs in terms of efficiency of cell collection by the unique swab matrix, which in conjunction with a quick release surface maximizes DNA/RNA yields. Swab batches are ethylene oxide treated and routinely tested for human DNA contamination.