Learn More
Andwin Scientific Tissue-Tek Feather Low Profile Blades, 500 pack (10 bags of 50)
SDP
Catalog No. NC1980065
Encompass
Tissue-Tek Feather Low Profile Blades, 50 pk, 0.24 (T) x 8 (H) x 80 (L) mm; Blade angle: 22° / mfg# 7089
Catalog No. NC1980065 Supplier Andwin Scientific Supplier No. 89199412
Tissue-Tek Feather Low Profile Blades, 50 pk, 0.24 (T) x 8 (H) x 80 (L) mm; Blade angle: 22° / mfg# 7089