Learn More
Research Products International Corp T Tubing Connectors, 1/4"", 12/pk
SDP
Catalog No. 501978376
Encompass
Connectors have multiple barbs per arm providing additional contact between the fitting and the tubing for extra gripping power. Ensures a tight, secure connection between lengths of tubing having the same inner diameter (I.D.). Chemical-resistant polypropylene for use with a wide range of liquids or gases. FDA-grade for direct contact with food, drugs, and cosmetics. Connectors are autoclavable.
Catalog No. 50-197-8376 Supplier Research Products International Corp Supplier No. F19604
Connectors have multiple barbs per arm providing additional contact between the fitting and the tubing for extra gripping power. Ensures a tight, secure connection between lengths of tubing having the same inner diameter (I.D.). Chemical-resistant polypropylene for use with a wide range of liquids or gases. FDA-grade for direct contact with food, drugs, and cosmetics. Connectors are autoclavable.