Learn More
Research Products International Corp T Shaped Tubing Connectors for 3/16 Inch Tubing, Polypropylene, 12 per Bag
SDP
Supplier:  Research Products International Corp F19603
Encompass
Connectors have multiple barbs per arm providing additional contact between the fitting and the tubing for extra gripping power. Ensures a tight, secure connection between lengths of tubing having the same inner diameter (I.D.). Chemical-resistant polypropylene for use with a wide range of liquids or gases. FDA-grade for direct contact with food, drugs, and cosmetics. Connectors are autoclavable.
Catalog No. 50-999-293