Learn More
Enzo Life Sciences T. vaginalis native antigen (1ml)
SDP
Supplier:  Enzo Life Sciences ALX6301301000
Encompass_Preferred
Alternative name: Trichomonas vaginalis native antigen. Purity: ≥99%. Appearance: Cloudy liquid. Formulation: Liquid. In sterile distilled water. Source: From Trichomonas vaginalis, strain C-1:NIH. Inactivated by freezing/thawing in water. Long Term Storage: -80°C. Applications: ELISA LINE assay, Immunogen, Cytotoxicity studies, Antibody capture.
Catalog No. 50-200-9655