Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza Walkersville TheraPEAK T-VIVO Cell Culture Medium, 5 L, Bag, Chemically Defined, Non-Animal Origin, Serum-Free, Contains Recombinant Proteins, For T cells, CAR-T, PBL, TIL

Catalog No. NC3771159
Encompass

Lonza Walkersville Inc.'s TheraPEAK T-VIVO Cell Culture Medium is a specifically formulated 5L bag for advanced hematopoietic cell culture applications. This serum-free, non-animal origin medium supports the expansion of various T cell subpopulations and is compliant with cGMP and ISO 13485 standards, ensuring high cell proliferation and process control. It simplifies regulatory compliance by eliminating the need for human serum and contains only recombinant proteins.

  • Volume: 5 L
  • Packaging: Bag
  • Formulation type: Chemically defined
  • Origin: Non-animal origin
  • Serum requirement: Serum-free
  • Applications: T cells, CAR-T cells, PBL, TIL
  • Quality standard: cGMP compliant
  • Scalability: Supports scale-up from preclinical to manufacturing
  • Storage: 2-8 °C, protected from light

This item is not returnable. View return policy
Catalog No. NC3771159 Supplier Lonza Walkersville Supplier No. BP08970H
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lonza Walkersville Inc.'s TheraPEAK T-VIVO Cell Culture Medium is a specifically formulated 5L bag for advanced hematopoietic cell culture applications. This serum-free, non-animal origin medium supports the expansion of various T cell subpopulations and is compliant with cGMP and ISO 13485 standards, ensuring high cell proliferation and process control. It simplifies regulatory compliance by eliminating the need for human serum and contains only recombinant proteins.

  • Volume: 5 L
  • Packaging: Bag
  • Formulation type: Chemically defined
  • Origin: Non-animal origin
  • Serum requirement: Serum-free
  • Applications: T cells, CAR-T cells, PBL, TIL
  • Quality standard: cGMP compliant
  • Scalability: Supports scale-up from preclinical to manufacturing
  • Storage: 2-8 °C, protected from light

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.