Learn More
Enzo Life Sciences T0070907 (50mg). CAS: 313516-66-4
SDP
Supplier:  Enzo Life Sciences BMLGR2120050
Encompass_Preferred
Potent and selective PPARγ antagonist. T0070907 blocks PPARγ function in both cell-based reporter gene and adipocyte differentiation assays. It inhibits growth and survival of hepatocellular carcinoma cells and squamous cell carcinoma cell lines by interfering with cell adhesion "anoikis". Alternative name: 2-Chloro-5-nitro-N-4-pyridinylbenzamide. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0935