Learn More
TARGETMOL CHEMICALS INC T0070907 50MG
SDP
Catalog No. 502579306
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. T0070907(IC50=1 nM) an effective and specific PPAR(gamma) inhibitor with the >800-fold selectivity over PPAR(alpha) and PPAR(delta). purity: 99%
Catalog No. 50-257-9306 Supplier TARGETMOL CHEMICALS INC Supplier No. T668950MG
Add to Cart
Add to Cart
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. T0070907(IC50=1 nM) an effective and specific PPAR(gamma) inhibitor with the >800-fold selectivity over PPAR(alpha) and PPAR(delta). purity: 99%