Learn More
Medchemexpress LLC HY-13202 5mg Medchemexpress, T0070907 CAS:313516-66-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY132025MG
Encompass_Preferred
Medchemexpress, HY-13202 5mg T0070907 CAS:313516-66-4 T0070907 is a potent PPARγ antagonist with a Ki of 1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1304