Learn More
Medchemexpress LLC T025 | 2407433-00-3 | 99.6% | 382.42 | 10 MG
SDP
Catalog No. 5000437222
Encompass_Preferred
T025 is an orally active and highly potent inhibitor of Cdc2-like kinase (CLKs). It induces caspase-3/7-mediated cell apoptosis, reduces CLK-dependent phosphorylation, and exhibits anti-proliferative activities in hematological and solid cancer cell lines. T025 primarily has anti-tumor efficiency for MYC-driven disease research.
  • Orally active and highly potent inhibitor of Cdc2-like kinase (CLKs)
  • Induces caspase-3/7-mediated cell apoptosis
  • Reduces CLK-dependent phosphorylation
  • Exhibits anti-proliferative activities in hematological and solid cancer cell lines
  • Anti-tumor efficiency for MYC-driven disease research
Catalog No. 50-004-37222 Supplier Medchemexpress LLC Supplier No. HY11229610MG
T025 is an orally active and highly potent inhibitor of Cdc2-like kinase (CLKs). It induces caspase-3/7-mediated cell apoptosis, reduces CLK-dependent phosphorylation, and exhibits anti-proliferative activities in hematological and solid cancer cell lines. T025 primarily has anti-tumor efficiency for MYC-driven disease research.
  • Orally active and highly potent inhibitor of Cdc2-like kinase (CLKs)
  • Induces caspase-3/7-mediated cell apoptosis
  • Reduces CLK-dependent phosphorylation
  • Exhibits anti-proliferative activities in hematological and solid cancer cell lines
  • Anti-tumor efficiency for MYC-driven disease research