Learn More
Medchemexpress LLC HY-112296 5mg Medchemexpress, T025 CAS: Purity:>98%
SDP
Catalog No. 501932419
Encompass_Preferred
Medchemexpress, HY-112296 5mg T025 CAS: T025 is an orally available and highly potent Cdc2-like kinase (CLK) inhibitor with K d s of 4.8, 0.096, 6.5, and 0.61 nM for CLK1, CLK2, CLK3, and CLK4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2419 Supplier Medchemexpress LLC Supplier No. HY1122965MG
Medchemexpress, HY-112296 5mg T025 CAS: T025 is an orally available and highly potent Cdc2-like kinase (CLK) inhibitor with K d s of 4.8, 0.096, 6.5, and 0.61 nM for CLK1, CLK2, CLK3, and CLK4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.