Learn More
Apexbio Technology LLC T0901317, 100mg. Cas: 293754-55-9 MFCD: MFCD03412028. Liver X receptor agonist,potent and selective
SDP
Supplier:  Apexbio Technology LLC A2249100
Encompass_Preferred
T0901317 is an agonist for multiple targets, which possesses EC50 values of 20 nM and 5 M for LXR and FXR, respectively. Furthermore, it is ROR and ROR dual inverse agonist with estimated IC50 of 2.0 M and 1.7 M, respectively. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4738