Learn More
Enzo Life Sciences T0901317 (50mg). CAS: 293754-55-9
SDP
Supplier:  Enzo Life Sciences ALX270309M050
Encompass_Preferred
Potent agonist (EC50=50nM) of the orphan nuclear hormone receptor (liver X receptor; LXR). Inhibitor of human sex hormone-dependent cancer cell lines. Alternative name: N-(2,2,2-trifluoroethyl)-N-[4-[2,2,2-trifluoro-1-hydroxy-1(trifluoromethyl)ethyl]phenyl]-benzenesulfonamide. Formula: C17H12NF9O3S. MW: 481.3. Purity: ≥98% (HPLC). Solubility: 50mg/ml soluble in 100% ethanol, DMSO, dimethyl formamide or any organic solvent; also soluble in a 1:1 solution of DMSO:PBS, pH 7.2 (500µg/ml). Use/Stability: We do not recommend storing the aqueous solutions for more than one day. Long Term Storage: -20°C.
Catalog No. 50-200-8482