Learn More
Abcam T0901317, L x R agonist, 10MG
Catalog No. 502706181
Encompass_Preferred
MW 481.3 Da, Purity >99%. Potent, non-selective LXR agonist (EC₅₀ = 50 nM). Increases ABCA1 expression associated with cholesterol efflux regulation and HDL metabolism. Increases muscle expression of PPAR-δ. Shows antiobesogenic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6181 Supplier Abcam Supplier No. AB14280810MG
MW 481.3 Da, Purity >99%. Potent, non-selective LXR agonist (EC₅₀ = 50 nM). Increases ABCA1 expression associated with cholesterol efflux regulation and HDL metabolism. Increases muscle expression of PPAR-δ. Shows antiobesogenic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement