Learn More
Alkali Scientific T150 U-Shaped Cell Culture Flask, Vented, Tissue Culture Treated
SDP
Catalog No. 503028277
Encompass_Preferred
A brand new specification, T150 U-shaped Canted Neck Cell Culture Flask, is added to the collection, which deviates from the regular design. Without influencing the regular usage, the bottleneck is enlarged, allowing the Pipettes or cell scrapers to reach every corner of the flask freely and flexibly. The unique U-shaped body is easier to hold and enhances the utilization rate of the bottom. A brand new specification, T150 U-shaped Canted Neck Cell Culture Flask, is added to the collection, which deviates from the regular design. Without influencing the regular usage, the bottleneck is enlarged, allowing the Pipettes or cell scrapers to reach every corner of the flask freely and flexibly. The unique U-shaped body is easier to hold and enhances the utilization rate of the bottom.
Catalog No. 50-302-8277 Supplier Alkali Scientific Supplier No. T150VTC
Add to Cart
Add to Cart
A brand new specification, T150 U-shaped Canted Neck Cell Culture Flask, is added to the collection, which deviates from the regular design. Without influencing the regular usage, the bottleneck is enlarged, allowing the Pipettes or cell scrapers to reach every corner of the flask freely and flexibly. The unique U-shaped body is easier to hold and enhances the utilization rate of the bottom. A brand new specification, T150 U-shaped Canted Neck Cell Culture Flask, is added to the collection, which deviates from the regular design. Without influencing the regular usage, the bottleneck is enlarged, allowing the Pipettes or cell scrapers to reach every corner of the flask freely and flexibly. The unique U-shaped body is easier to hold and enhances the utilization rate of the bottom.