Learn More
Alkali Scientific T150 U-Shaped Cell Culture Flask, Vented, Non-Treated
SDP
Catalog No. 503027003
Encompass_Preferred
A brand new specification, T150 U-shaped Canted Neck Cell Culture Flask, is added to the collection, which deviates from the regular design. Without influencing the regular usage, the bottleneck is enlarged, allowing the Pipettes or cell scrapers to reach every corner of the flask freely and flexibly. The unique U-shaped body is easier to hold and enhances the utilization rate of the bottom. A brand new specification, T150 U-shaped Canted Neck Cell Culture Flask, is added to the collection, which deviates from the regular design. Without influencing the regular usage, the bottleneck is enlarged, allowing the Pipettes or cell scrapers to reach every corner of the flask freely and flexibly. The unique U-shaped body is easier to hold and enhances the utilization rate of the bottom.
Catalog No. 50-302-7003 Supplier Alkali Scientific Supplier No. T150VNT
A brand new specification, T150 U-shaped Canted Neck Cell Culture Flask, is added to the collection, which deviates from the regular design. Without influencing the regular usage, the bottleneck is enlarged, allowing the Pipettes or cell scrapers to reach every corner of the flask freely and flexibly. The unique U-shaped body is easier to hold and enhances the utilization rate of the bottom. A brand new specification, T150 U-shaped Canted Neck Cell Culture Flask, is added to the collection, which deviates from the regular design. Without influencing the regular usage, the bottleneck is enlarged, allowing the Pipettes or cell scrapers to reach every corner of the flask freely and flexibly. The unique U-shaped body is easier to hold and enhances the utilization rate of the bottom.