Learn More
Cayman Chemical T16A Inh-A01 10mg
SDP
Catalog No. 502967899
Encompass_Preferred
An aminophenylthiazole that inhibits transient TMEM16A-mediated chloride currents with an IC50 value of ~1 µM; blocks CaCC activity in vascular smooth muscle cells and relaxes mouse and human blood vessels
Catalog No. 50-296-7899 Supplier Cayman Chemical Supplier No. 1851810 MG
Add to Cart
Add to Cart
An aminophenylthiazole that inhibits transient TMEM16A-mediated chloride currents with an IC50 value of ~1 µM; blocks CaCC activity in vascular smooth muscle cells and relaxes mouse and human blood vessels