Learn More
Cayman Chemical T16A(inh)-A01, Purity: ≥95%, CAS: 552309-42-9, 50 mg
SDP
Catalog No. 502965257
Encompass_Preferred
An aminophenylthiazole that inhibits transient TMEM16A-mediated chloride currents with an IC50 value of ~1 µM; blocks CaCC activity in vascular smooth muscle cells and relaxes mouse and human blood vessels
Catalog No. 50-296-5257 Supplier Cayman Chemical Supplier No. 1851850 MG
An aminophenylthiazole that inhibits transient TMEM16A-mediated chloride currents with an IC50 value of ~1 µM; blocks CaCC activity in vascular smooth muscle cells and relaxes mouse and human blood vessels